Learn More
TARGETMOL CHEMICALS INC Durlobactam sodium salt 1MG
SDP
Catalog No. 502879496
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Durlobactam sodium salt (ETX2514) is a beta-lactamase inhibitor with IC50 values of 4 nM, 14 nM and 190 nM against class A KPC-2, class C AmpC and class D OXA-24. Purity 99.03%
Catalog No. 50-287-9496 Supplier TARGETMOL CHEMICALS INC Supplier No. T111251MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Durlobactam sodium salt (ETX2514) is a beta-lactamase inhibitor with IC50 values of 4 nM, 14 nM and 190 nM against class A KPC-2, class C AmpC and class D OXA-24. Purity 99.03%