Learn More
MSC Mitutoyo Hardness Calibration Set, Shore D, 3 Piece, 1 ea
Catalog No. 5000512319
Encompass
This hardness test block set is designed for the calibration and verification of Shore D durometers, specifically for use with Hardmatic HH-300 models. The set includes three reference blocks to ensure accurate hardness testing measurements across the specified Shore D range.
  • Includes three reference blocks with nominal values of 20, 40, and 80
  • Calibration set specifically for the Shore D hardness scale
  • Provides reliable reference standards for durometer testing
  • Designed for use with Hardmatic HH-300 instrumentation
Catalog No. 50-005-12319 Supplier MSC Supplier No. 39059019
This hardness test block set is designed for the calibration and verification of Shore D durometers, specifically for use with Hardmatic HH-300 models. The set includes three reference blocks to ensure accurate hardness testing measurements across the specified Shore D range.
  • Includes three reference blocks with nominal values of 20, 40, and 80
  • Calibration set specifically for the Shore D hardness scale
  • Provides reliable reference standards for durometer testing
  • Designed for use with Hardmatic HH-300 instrumentation