Description
- Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose.
- Smooth, highly absorbent and durable.
- Low extractable levels and particle counts.
- Chemically compatible with common cleaning and disinfecting solutions.
- Autoclavable.
Specifications
Specifications
| Product Type | Absorbent Wipe |
| Material | Polyester Cellulose (Binder-free) |
| Autoclavable | Autoclavable |
| Disposable | Single-use |
| Ply | 1-ply |
| Sterility | Non-sterile |