Learn More
Aviva Systems Biology Corp DUS1L Antibody - middle region (ARP57616_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57616_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DUS1L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DUS1L. Alternate names: DUS1, PP3111ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-62912