Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Dusan™

Catalog No. 17000056
Change view
Click to view available options
Volume (Metric):
3.78 L
208 L
Quantity:
1 Drum
4/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity
17-000-056 3.78 L 4/Cs.
17-000-055 208 L 1 Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-000-056 Supplier Diversey™ Supplier No. 1833276
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hard surface sanitizer

  • Acidic low foaming iodophor

Specifications

Product Line Dusan
Sterility Non-sterile
Appearance Brown
Certifications/Compliance EPA, Kosher
For Use With (Application) Sanitizing hard surfaces in the food service, healthcare and allied industries
Quantity 4/Cs.
Volume (English) 1 gal.
Volume (Metric) 3.78 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.