Learn More
Aviva Systems Biology Corp DUSP10 Antibody - N-terminal region (ARP52762_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52762_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DUSP10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DUSP10. Alternate names: MKP-5, MKP5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60825