Learn More
Sino Biological DUSP14 / MKP-6 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 12443RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DUSP14 / MKP-6 (rh DUSP14; Catalog#12443-H10E; O95147; Met1-His191). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-7953