Learn More
Sino Biological DUSP14 / MKP-6 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 12443T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DUSP14 / MKP-6 (rh DUSP14 / MKP-6; Catalog#12443-H10E; O95147; Met1-His191). DUSP14 / MKP-6 specific IgG was purified by Human DUSP14 / MKP-6 affinity chromatography.
Catalog No. 50-224-7956