Learn More
Aviva Systems Biology Corp DUSP19 Antibody - C-terminal region (ARP52531_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52531_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DUSP19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DUSP19. Alternate names: DUSP17, MGC138210, SKRP1, TS-DSP1, LMWDSP3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60763