Learn More
Eurofins Discoverx DUSP22, 10 µg. (14-641)
Supplier:  Eurofins Discoverx 14641
Encompass
The DUSP22 product is purified N terminal GST-tagged recombinant DUSP22 enzyme. This enzyme is full length. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 10 μg of 1.629 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9103