Learn More
Enzo Life Sciences VHR dual specificity phosphatase (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE3330050
Encompass_Preferred
Alternative name: Dual specificity protein phosphatase 3, DUSP3. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM citrate, pH 6.0, containing 0.1M sodium chloride, 1mM EDTA and 1mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-1987