Learn More
Aviva Systems Biology Corp DUSP4 Antibody - N-terminal region (ARP64084_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64084_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DUSP4. It was validated on Western Blot. . Alternate names: HVH2, MKP-2, MKP2, TYPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-169