Learn More
Aviva Systems Biology Corp DUSP5 Antibody - middle region (ARP58225_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58225_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DUSP5. It was validated on Western Blot. DUSP5,dual specificity phosphatase 5,DUSP; HVH3; DUSP5,dual specificity protein phosphatase 5,dual specificity protein phosphatase 5,VH1-like phosphatase 3,dual specificity protein phosphatase hVH3,serine/threonine specific protein phosphataseThe immunogen is a synthetic peptide directed towards the middle region of human DUSP5. Alternate names: DUSP, HVH3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-453