Learn More
Aviva Systems Biology Corp DUSP9 Antibody - middle region (ARP61993_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61993_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DUSP9. It was validated on Western Blot. . Alternate names: MKP-4, MKP4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-287-535