Learn More
MSC Soilworks Soiltac Sample, Biodegradable, Dries Transparent, 1 lb
Catalog No. 502839668
Encompass
Soiltac is an advanced copolymer emulsion designed for durable dust control, soil stabilization, and erosion mitigation. It forms strong, water-resistant bonds, creating a flexible, transparent matrix that stabilizes soil while preserving natural appearance. This eco-safe, biodegradable, liquid copolymer effectively links and cross-links soil particles into a durable, water-resistant solid mass.
  • Dries transparent/clear
  • Non-hazardous and non-toxic
  • Biodegradable and eco-safe
  • Water resistant
  • Non-flammable and non-volatile
  • Safe for humans, animals, and vegetation
  • PM10 and PM2.5 compliant
Catalog No. 50-283-9668 Supplier MSC Supplier No. 78986106
Add to Cart
Add to Cart
Request a Quote
Soiltac is an advanced copolymer emulsion designed for durable dust control, soil stabilization, and erosion mitigation. It forms strong, water-resistant bonds, creating a flexible, transparent matrix that stabilizes soil while preserving natural appearance. This eco-safe, biodegradable, liquid copolymer effectively links and cross-links soil particles into a durable, water-resistant solid mass.
  • Dries transparent/clear
  • Non-hazardous and non-toxic
  • Biodegradable and eco-safe
  • Water resistant
  • Non-flammable and non-volatile
  • Safe for humans, animals, and vegetation
  • PM10 and PM2.5 compliant