Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Dust Cover for SP12 Soft-Flo Heads
SDP

Catalog No. 19033736 Shop All Haws Corporation Products
Change view
Click to view available options
Material:
Polyethylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-033-736 Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-033-736 Supplier Haws™ Supplier No. 9094
  • $73.75
    $54.65 / Pack of 2

    Save $19.10

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keeps eyewash heads from contamination when not in use.

  • For use on all Haws Portable Eyewash Units that feature the SP12 Soft-Flo heads
  • Automatically releases when water is turned on
  • Green
  • Polyethylene
  • With integral retainer strap
  • Sold in pairs

Specifications

Color Green
For Use With (Equipment) Haws Portable Eyewash Units that feature the SP12 Soft-Flo heads.
Includes Integral Retainer Strap
Material Polyethylene
Product Type Eyewash Dust Cover
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.