Learn More
Electron Microscopy Sciences Dust-Off DPN Kit
SDP
Catalog No. 50930498
Encompass_Preferred
(Polyethylene glycol tert-octylphenyl ether)
Contains: Formula N (Tetrafluoroethane) - 100% safe for the ozone. The same as DPSA above but this formula-as mentioned-contains no flammable elements. Kit consists of a 8 oz can and a chrome nozzle.
Catalog No. 50-930-498 Supplier Electron Microscopy Sciences Supplier No. 70846
(Polyethylene glycol tert-octylphenyl ether)
Contains: Formula N (Tetrafluoroethane) - 100% safe for the ozone. The same as DPSA above but this formula-as mentioned-contains no flammable elements. Kit consists of a 8 oz can and a chrome nozzle.