Learn More
Electron Microscopy Sciences Dust-Off Fg Chrome Nozzle Only
SDP
Supplier:  Electron Microscopy Sciences 70702
Encompass_Preferred
Contains Formula S (Difluoroethane). It is 100% safe for the ozone with no CFC's, no HCFC's, no propane, and no butane. Considered non-flammable (Formula S), and completely safe for normal use, however this formula is considered flammable for shipping purposes and does contain flammable elements.
The chrome nozzle is designed to prevent droplets from forming. Delivers pure, moisture-free blasts which removes all dust and dirt particles. It retains the same pressure until the can is empty. The kit consists of a 8 oz. can and a chrome nozzle. The nozzle is designed to prevent droplets from forming, use with Dust-Off FGSA.
Catalog No. 50-930-481