Learn More
Electron Microscopy Sciences Dust-Off FGSA Refill 8 OZ
SDP
Catalog No. 50930484
Encompass_Preferred
Contains Formula S (Difluoroethane). It is 100% safe for the ozone with no CFC's, no HCFC's, no propane, and no butane. Considered non-flammable (Formula S), and completely safe for normal use, however this formula is considered flammable for shipping purposes and does contain flammable elements.
The chrome nozzle is designed to prevent droplets from forming. Delivers pure, moisture-free blasts which removes all dust and dirt particles. It retains the same pressure until the can is empty. The kit consists of a 8 oz. can and a chrome nozzle. Contains: Formula S (Difluoroethane) - 100% safe for the ozone.
Catalog No. 50-930-484 Supplier Electron Microscopy Sciences Supplier No. 70707
Contains Formula S (Difluoroethane). It is 100% safe for the ozone with no CFC's, no HCFC's, no propane, and no butane. Considered non-flammable (Formula S), and completely safe for normal use, however this formula is considered flammable for shipping purposes and does contain flammable elements.
The chrome nozzle is designed to prevent droplets from forming. Delivers pure, moisture-free blasts which removes all dust and dirt particles. It retains the same pressure until the can is empty. The kit consists of a 8 oz. can and a chrome nozzle. Contains: Formula S (Difluoroethane) - 100% safe for the ozone.