Learn More
Electron Microscopy Sciences Dust-Off Plus Kit(DPSA)
SDP
Catalog No. 50930492
Encompass_Preferred
Contains: Formula S (Difluoroethane) - 100% safe for the ozone. It comes with a patented 360 degree Vector Valve. The multi-directional valve is used for easy blasting in hard to reach areas. The valve can be removed from the can anytime without any loss of gas. Its new concentrated formula will give you 57% more blasts per ounce, 17% more blasts per can. The kit consists of a 8 oz. can and a Vector Valve nozzle.
Catalog No. 50-930-492 Supplier Electron Microscopy Sciences Supplier No. 70840
Contains: Formula S (Difluoroethane) - 100% safe for the ozone. It comes with a patented 360 degree Vector Valve. The multi-directional valve is used for easy blasting in hard to reach areas. The valve can be removed from the can anytime without any loss of gas. Its new concentrated formula will give you 57% more blasts per ounce, 17% more blasts per can. The kit consists of a 8 oz. can and a Vector Valve nozzle.