Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andwin Scientific Dust-Off™ Plus Pressurized Duster
SDP

Catalog No. 1437364
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-373-64 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-373-64 Supplier Andwin Scientific Supplier No. 21899090

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Removes dust, lint, and more from computers, CRT's, disk and tape drives, and general office equipment

  • 8 oz. (227g) can with 360-degree Vector Valve
  • Nonflammable, clean, nontoxic gas - ozone safe; no CFCs

Specifications

Product Type Pressurized Duster
Flammable No
For Use With (Application) Removes dust, lint and more from computers, CRT's, disk and tape drives and general office equipment
Quantity 8 oz.
Volume (English) 8 oz.
Volume (Metric) 227 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.