Learn More
Electron Microscopy Sciences Dust-Off Plus Refill(DPSA) 8 OZ
SDP
Catalog No. 50930493
Encompass_Preferred
Contains: Formula S (Difluoroethane) - 100% safe for the ozone. It comes with a patented 360 degree Vector Valve. The multi-directional valve is used for easy blasting in hard to reach areas. The valve can be removed from the can anytime without any loss of gas. Its new concentrated formula will give you 57% more blasts per ounce, 17% more blasts per can.
Catalog No. 50-930-493 Supplier Electron Microscopy Sciences Supplier No. 70843
Contains: Formula S (Difluoroethane) - 100% safe for the ozone. It comes with a patented 360 degree Vector Valve. The multi-directional valve is used for easy blasting in hard to reach areas. The valve can be removed from the can anytime without any loss of gas. Its new concentrated formula will give you 57% more blasts per ounce, 17% more blasts per can.