Learn More
Ted Pella Inc Dust-Off Plus Valve only, each
SDP
Catalog No. NC1694504
Encompass
Dust-off general purpose cleaner with difluoroethane as active gas. Widely used in microscopy labs. The unique Dust-Off multi directional nozzle can be angled and rotated for optimum delivery of the cleaning gas. Ideal for cleaning microscopes, precision instruments, computer parts, samples etc.
Catalog No. NC1694504 Supplier Ted Pella Inc Supplier No. 81504
Dust-off general purpose cleaner with difluoroethane as active gas. Widely used in microscopy labs. The unique Dust-Off multi directional nozzle can be angled and rotated for optimum delivery of the cleaning gas. Ideal for cleaning microscopes, precision instruments, computer parts, samples etc.