Learn More
Ted Pella Inc DUST OFF REFILL/EACH
SDP
Catalog No. NC9152917
Encompass
Dust-Off Plus Refills, 10 oz., Pressurized cleaner w/multidirectional nozzle, no CFC's, HCFC's, Dusters, Lint remover, Flammable: 100% Ozone-Safe, Boiling Point: -40.8degreeC (-41.4degreeF), Vapor Density (Air: 1): 3.03 at 25degreeC(77degre
Catalog No. NC9152917 Supplier Ted Pella Inc Supplier No. 81505
Add to Cart
Add to Cart
Dust-Off Plus Refills, 10 oz., Pressurized cleaner w/multidirectional nozzle, no CFC's, HCFC's, Dusters, Lint remover, Flammable: 100% Ozone-Safe, Boiling Point: -40.8degreeC (-41.4degreeF), Vapor Density (Air: 1): 3.03 at 25degreeC(77degre