Learn More
Andwin Scientific DUST-OFF XL (DPSXL)12/CSX10 OZ
SDP
Catalog No. 50930491
Encompass_Preferred
The Dust-Off XL is an effective solution for removing dust and contaminants with pure moisture free blasts. This product contains Formula S or Difluoroethane, which is 100% safe for the ozone. It is packaged in a 10 oz. can and is sold in a case of 12. This product also includes an extension tube.
Catalog No. 50-930-491 Supplier Andwin Scientific Supplier No. 70838
The Dust-Off XL is an effective solution for removing dust and contaminants with pure moisture free blasts. This product contains Formula S or Difluoroethane, which is 100% safe for the ozone. It is packaged in a 10 oz. can and is sold in a case of 12. This product also includes an extension tube.