Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Ampco Handheld Dustpan, Aluminum Body, 5 in Handle, 1 ea

Catalog No. 502817779
Encompass

This handheld dustpan features an aluminum body, designed for durability and safety in various environments. It offers corrosion resistance, non-magnetic properties, and non-sparking characteristics, meeting FM standards for hazardous applications.

  • Corrosion resistant
  • Non-magnetic
  • Non-sparking
  • Meets FM standards
  • Handheld type
  • Aluminum body construction
  • Handle length: 5 in
  • Pan length: 4 in
  • Pan width: 4 in
  • Depth: 13 in
  • Height: 1-1/2 in

Catalog No. 50-281-7779 Supplier MSC Supplier No. 05569942
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This handheld dustpan features an aluminum body, designed for durability and safety in various environments. It offers corrosion resistance, non-magnetic properties, and non-sparking characteristics, meeting FM standards for hazardous applications.

  • Corrosion resistant
  • Non-magnetic
  • Non-sparking
  • Meets FM standards
  • Handheld type
  • Aluminum body construction
  • Handle length: 5 in
  • Pan length: 4 in
  • Pan width: 4 in
  • Depth: 13 in
  • Height: 1-1/2 in

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.