Learn More
Electron Microscopy Sciences Dust-Pro Kit
SDP
Catalog No. 50930495
Encompass_Preferred
For dusting and chilling 100% ozone-safe, pressurized Tetrafluoroethane. Triple filtered for maximum purity. The one-piece-valve is made from solid brass and is chrome plated with an extra long nozzle (3 1/2") for hard-to-reach areas. No mounting clip is required. The valve can be removed at any time without loss of pressure. Meets Canadian Government Regulation. Kit includes one 8 oz. can Dust-Pro and Peca Techni-Valve.
Catalog No. 50-930-495 Supplier Electron Microscopy Sciences Supplier No. 7084500
For dusting and chilling 100% ozone-safe, pressurized Tetrafluoroethane. Triple filtered for maximum purity. The one-piece-valve is made from solid brass and is chrome plated with an extra long nozzle (3 1/2") for hard-to-reach areas. No mounting clip is required. The valve can be removed at any time without loss of pressure. Meets Canadian Government Regulation. Kit includes one 8 oz. can Dust-Pro and Peca Techni-Valve.