Learn More
ACL Inc ACL Turbo Blast, 11 ounce, 12/cs
Catalog No. 501037863
Encompass_Preferred
ESD workbench products, air duster, aCL Staticide, Staticide Turbo Blast, precision air duster, canned air, exceptional purity. Economical High Precision Duster rids surfaces of particulate contaminates without leaving residuals. Engineered to remove dirt, dust, oxide particles. Excellent in using to removing entrapped solvents from hard to reach area, as well accelerating drying time.
Catalog No. 50-103-7863 Supplier ACL Inc Supplier No. 8640
ESD workbench products, air duster, aCL Staticide, Staticide Turbo Blast, precision air duster, canned air, exceptional purity. Economical High Precision Duster rids surfaces of particulate contaminates without leaving residuals. Engineered to remove dirt, dust, oxide particles. Excellent in using to removing entrapped solvents from hard to reach area, as well accelerating drying time.