Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Dustless Precision Pellets™ Grain-Based Rodent Diet
SDP

Catalog No. 14726587
Encompass
Change view
Click to view available options
Quantity:
0.02 g
0.045 g
0.5 g
1 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-726-587 1 g
14-726-581 0.02 g
14-726-582 0.045 g
14-726-585 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-726-587 Supplier Bio-Serv™ Supplier No. F0173
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Precise size and weight pellets generally used in nutritional and behavioral studies

  • Nutritionally complete grain-based formula
  • Dustless and fracture free—superior performance in automatic feeders
  • Every batch tested for hardness, weight and height
  • Nutritionally assayed

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin

Specifications

Product Type Rodent Diet
Calories 3.35
Components Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin
Format Pellet
For Use With (Application) Nutritional and behavioral studies
Particle Shape Pellet
Quantity 1 g
Species Rodent
Storage Requirements Cool dry place
Diameter (English) 0.45 in.
Diameter (Metric) 0.114 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.