Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Dustless Precision Pellets™ Purified Rodent Diet
SDP

Catalog No. 14726750
Encompass
Change view
Click to view available options
Diameter (English):
0.1 in.
0.14 in.
0.3 in.
Diameter (Metric):
0.025 cm
0.036 cm
0.076 cm
Quantity:
0.02 g
0.045 g
0.3 g
Flavor:
Chocolate
No Added Flavor
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Diameter (Metric) Quantity Flavor
14-726-750 0.14 in. 0.036 cm 0.045 g No Added Flavor
14-726-596 0.14 in. 0.036 cm 0.045 g Chocolate
14-726-595 0.1 in. 0.025 cm 0.02 g Chocolate
14-726-589 0.1 in. 0.025 cm 0.02 g No Added Flavor
14-726-592 0.3 in. 0.076 cm 0.3 g No Added Flavor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-726-750 Supplier Bio-Serv™ Supplier No. F0021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Precise size and weight pellets generally used in nutritional and behavioral studies

Precise size and weight pellets generally used in nutritional and behavioral studies

  • Nutritionally complete purified formula
  • Dustless and fracture free—superior performance in automatic feeders
  • Every batch tested for hardness, weight and height
  • Nutritionally assayed

Ingredients: Sucrose. Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ, Chocolate Flavoring (F05301 and F0299 only)

Specifications

Product Type Rodent Diet
Calories 3.6
Components Sucrose, Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ
Flavor No Added Flavor
Format Pellet
For Use With (Application) Nutritional and behavioral studies
Particle Shape Pellet
Quantity 0.045 g
Species Rodent
Storage Requirements Cool dry place
Diameter (English) 0.14 in.
Diameter (Metric) 0.036 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.