Learn More
Aviva Systems Biology Corp DUT Antibody - N-terminal region (ARP46028_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46028_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DUT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DUT. Alternate names: FLJ20622, dUTPaseApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-59183