Learn More
Medchemexpress LLC HY-13613 10mg Medchemexpress, Dutasteride CAS:164656-23-9 Purity:>98%
SDP
Catalog No. 502081422
Encompass_Preferred
Medchemexpress, HY-13613 10mg Dutasteride CAS:164656-23-9 Dutasteride (GG745) is a potent inhibitor of both 5α-reductase isozymes. Dutasteride may possess off-target effects on the androgen receptor (AR) due to its structural similarity to DHT. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1422 Supplier Medchemexpress LLC Supplier No. HY1361310MG
Medchemexpress, HY-13613 10mg Dutasteride CAS:164656-23-9 Dutasteride (GG745) is a potent inhibitor of both 5α-reductase isozymes. Dutasteride may possess off-target effects on the androgen receptor (AR) due to its structural similarity to DHT. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.