Learn More
Selleck Chemical LLC Dutasteride 10mg 164656-23-9 GI198745, GG-745
SDP
Catalog No. 506246
Encompass_Preferred
Dutasteride (GI198745, GG-745) is a dual 5-? reductase inhibitor that inhibits conversion of testosterone to dihydrotestosterone (DHT). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-624-6 Supplier Selleck Chemical LLC Supplier No. S120210MG
Dutasteride (GI198745, GG-745) is a dual 5-? reductase inhibitor that inhibits conversion of testosterone to dihydrotestosterone (DHT). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.