Learn More
Sigma Aldrich Fine Chemicals Biosciences dUTP PCR Grade sodium salt
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 3732720001
Encompass
dUTP PCR Grade is a 100 mM clear colorless SOLUTIONon of the sodium salt (pH 8.3).
Catalog No. 50-291-4648