Learn More
Sigma Aldrich Fine Chemicals Biosciences dUTP PCR Grade sodium salt
Catalog No. 502903794
Encompass
dUTP PCR Grade is a 100 mM clear colorless SOLUTIONon of the sodium salt (pH 8.3).
Catalog No. 50-290-3794 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11969056001
dUTP PCR Grade is a 100 mM clear colorless SOLUTIONon of the sodium salt (pH 8.3).