Learn More
Medchemexpress LLC HY-17044 10mg , Duvelisib CAS:1201438-56-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1704410MG
Encompass_Preferred
HY-17044 10mg Duvelisib CAS: 1201438-56-3 Duvelisib (IPI-145) is a selectivite p100δ inhibitor with IC50 of 2.5 nM, 27.4 nM, 85 nM and 1602 nM for p110δ, P110γ, p110β and p110α, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0996