Learn More
Selleck Chemical LLC Duvelisib
SDP
Catalog No. 5000002129
Encompass_Preferred
Duvelisib (IPI-145 INK1197) is a novel and selective PI3K / inhibitor with Ki and IC50 of 23 pM/243 pM and 1 nM/50 nM in cell-free assays highly selective for PI3K / than other protein kinases Phase 3
Catalog No. 50-000-02129 Supplier Selleck Chemical LLC Supplier No. S702850MG
Duvelisib (IPI-145 INK1197) is a novel and selective PI3K / inhibitor with Ki and IC50 of 23 pM/243 pM and 1 nM/50 nM in cell-free assays highly selective for PI3K / than other protein kinases Phase 3