Learn More
Enzo Life Sciences Duvelisib (free base) (200mg). CAS: 1201438-56-3
SDP
Catalog No. 502013596
Encompass_Preferred
Duvelisib is an orally bioavailable and highly selective inhibitor of PI3K-δ and PI3K-γ with IC50 values of 0.36 nM and 19.6 nM, respectively. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3596 Supplier Enzo Life Sciences Supplier No. ENZCHM2230200
Duvelisib is an orally bioavailable and highly selective inhibitor of PI3K-δ and PI3K-γ with IC50 values of 0.36 nM and 19.6 nM, respectively. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.