Learn More
Apexbio Technology LLC Duvelisib (IPI-145, INK1197), 5mg. Cas: 1201438-56-3 MFCD: MFCD15144635. PI3K-δ/PI3K-γ inhibitor
SDP
Supplier:  Apexbio Technology LLC A17205
Encompass_Preferred
IPI-145 (also known as INK1197), an isoquinolinone derivative, is a small-molecule phosphoinositide-3 kinases (PI3K) inhibitor that selectively and potently inhibits two PI3K isoforms, PI3K- and PI3K-, with half maximal inhibition concentration IC50 of 2.5 nM and 27 nM respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4705