Learn More
Aviva Systems Biology Corp Duxbl Antibody - middle region (ARP37609_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37609_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Duxbl. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1110051B16Rik, Duxl, MGC67567, DuxblApplicationWB. Species reactivity: Mouse
Catalog No. 50-284-669