Learn More
Medchemexpress LLC DV1 TFA | 98.5% | 2424.85 | 1 MG
SDP
Catalog No. 5000423698
Encompass_Preferred
DV1 TFA is a selective antagonist of CXC chemokine receptor 4 (CXCR4). It exhibits antiviral activity by blocking HIV-1 entry through the CXCR4 co-receptor.
  • Selective antagonist for CXC chemokine receptor 4 (CXCR4)
  • Exhibits antiviral activity
  • Blocks HIV-1 entry through the CXCR4 co-receptor
Catalog No. 50-004-23698 Supplier Medchemexpress LLC Supplier No. HYP10427A1MG
DV1 TFA is a selective antagonist of CXC chemokine receptor 4 (CXCR4). It exhibits antiviral activity by blocking HIV-1 entry through the CXCR4 co-receptor.
  • Selective antagonist for CXC chemokine receptor 4 (CXCR4)
  • Exhibits antiviral activity
  • Blocks HIV-1 entry through the CXCR4 co-receptor