Learn More
Medchemexpress LLC DV1 TFA | 98.53% | 2424.85 | 25 MG
SDP
Catalog No. 5000426798
Encompass_Preferred
DV1 TFA is a selective antagonist for CXC chemokine receptor 4 (CXCR4). It exhibits antiviral activity by blocking HIV-1 entry through the CXCR4 co-receptor, making it suitable for research applications.
  • Selective antagonist for CXC chemokine receptor 4 (CXCR4)
  • Exhibits antiviral activity
  • Blocks HIV-1 entry
Catalog No. 50-004-26798 Supplier Medchemexpress LLC Supplier No. 25MGHYP10427A
DV1 TFA is a selective antagonist for CXC chemokine receptor 4 (CXCR4). It exhibits antiviral activity by blocking HIV-1 entry through the CXCR4 co-receptor, making it suitable for research applications.
  • Selective antagonist for CXC chemokine receptor 4 (CXCR4)
  • Exhibits antiviral activity
  • Blocks HIV-1 entry