Learn More
Brookfield Engineer Labs AMETEK Brookfield DV2Plus Viscometer, 740 Speeds, 7" Touchscreen, 1 ea
Catalog No. NC3939103
Encompass
The DV2Plus Viscometer is a high-precision digital instrument designed for comprehensive viscosity measurement, ranging from thin inks to heavy industrial pastes. Featuring a 7-inch full-color touchscreen, it provides intuitive operation and real-time data monitoring for applications across quality control and research and development.
  • Accuracy of ±1.0% and repeatability of ±0.2% of range.
  • Seven-inch full-color PCAP touchscreen with 800x480 resolution.
  • Extensive speed control with 740 user-selectable speeds.
  • Integrated gel timer and automated oscillation testing capabilities.
  • Comprehensive connectivity including four USB-A ports, one USB-B port, and Bluetooth.
  • Digital leveling function for simplified setup and maintenance.
  • Support for viscosity, temperature, shear rate, and torque measurement.
Catalog No. NC3939103 Supplier Brookfield Engineer Labs Supplier No. XDV2PHBTBB0U00
The DV2Plus Viscometer is a high-precision digital instrument designed for comprehensive viscosity measurement, ranging from thin inks to heavy industrial pastes. Featuring a 7-inch full-color touchscreen, it provides intuitive operation and real-time data monitoring for applications across quality control and research and development.
  • Accuracy of ±1.0% and repeatability of ±0.2% of range.
  • Seven-inch full-color PCAP touchscreen with 800x480 resolution.
  • Extensive speed control with 740 user-selectable speeds.
  • Integrated gel timer and automated oscillation testing capabilities.
  • Comprehensive connectivity including four USB-A ports, one USB-B port, and Bluetooth.
  • Digital leveling function for simplified setup and maintenance.
  • Support for viscosity, temperature, shear rate, and torque measurement.