Learn More
Grainger DVI Coupler (F to F)
Catalog No. 502748958
Encompass_Preferred
The Monoprice Video Adapter Adapter is a passive VGA to HDMI converter that allows you to connect a VGA source device such as a computer to an HDMI display such as a monitor or projector. It supports resolutions up to 1920x1200 and 1080p. The adapter does not require any external power source. It has 15-pin HD-15 VGA male input and 19-pin HDMI female output connectors. The adapter measures 5 inches long and weighs 0.8 ounces. It has gold-plated connectors and molded cables for maximum conductivity and durability. The adapter works with PCs, laptops, Blu-ray players, and other devices with VGA output. It comes with a 1 year warranty from Monoprice.
Catalog No. 50-274-8958 Supplier Grainger Supplier No. 14C356
The Monoprice Video Adapter Adapter is a passive VGA to HDMI converter that allows you to connect a VGA source device such as a computer to an HDMI display such as a monitor or projector. It supports resolutions up to 1920x1200 and 1080p. The adapter does not require any external power source. It has 15-pin HD-15 VGA male input and 19-pin HDMI female output connectors. The adapter measures 5 inches long and weighs 0.8 ounces. It has gold-plated connectors and molded cables for maximum conductivity and durability. The adapter works with PCs, laptops, Blu-ray players, and other devices with VGA output. It comes with a 1 year warranty from Monoprice.