Learn More
Aviva Systems Biology Corp DVL1 Antibody - C-terminal region (ARP32101_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32101_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DVL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DVL1. Alternate names: DVL, MGC54245, DVL1L1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-54451