Learn More
Aviva Systems Biology Corp DVL1 Antibody - middle region (ARP32100_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32100_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DVL1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human DVL1. Alternate names: DVL, MGC54245, DVL1L1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54450