Learn More
Hach Company Inorganics Quality Control Standard for Drinking Water, 500 mL
Supplier:  Hach Company 2833049
Encompass
Inorganics Quality Control Standard for Drinking Water. Ready to use. No dilution required. Mixed standard includes: Fluoride: 1 mg/L F⁻ , Nitrate: 2 mg/L NO₃⁻-N, Phosphate: 2 mg/L PO₄³⁻, Sulfate: 50 mg/L SO₄²⁻. 500 mL Bottle.
Catalog No. 50-199-6735