Learn More
Medchemexpress LLC DW71177 | 2241311-72-6 | 97.7% | 384.48 | 50 MG
SDP
Catalog No. 5000394959
Encompass_Preferred
DW71177 is a novel triazolo[4,3-a] quinoxaline-based potent and BD1-Selective BET inhibitor. It can be used for the study of acute myeloid leukemia.
  • Novel triazolo[4,3-a] quinoxaline-based compound
  • Potent and BD1-selective BET inhibitor
  • Can be used for the study of acute myeloid leukemia
Catalog No. 50-003-94959 Supplier Medchemexpress LLC Supplier No. HY16208850MG
DW71177 is a novel triazolo[4,3-a] quinoxaline-based potent and BD1-Selective BET inhibitor. It can be used for the study of acute myeloid leukemia.
  • Novel triazolo[4,3-a] quinoxaline-based compound
  • Potent and BD1-selective BET inhibitor
  • Can be used for the study of acute myeloid leukemia