Learn More
Electron Microscopy Sciences EMS High Alloy DX Biological Tweezers Style 3, 120 mm OAL, 0.08 mm Tip width
SDP
Catalog No. 501930610
Encompass_Preferred
The high alloy biological tweezers are anti-acid and anti-magnetic. The handles are serrated. Tips: Straight, very sharp, fine, superior finish.
OAL: 120 mm
Tip width: 0.08 mm
Tip thickness: 0.04 mm
Catalog No. 50-193-0610 Supplier Electron Microscopy Sciences Supplier No. 785113
The high alloy biological tweezers are anti-acid and anti-magnetic. The handles are serrated. Tips: Straight, very sharp, fine, superior finish.
OAL: 120 mm
Tip width: 0.08 mm
Tip thickness: 0.04 mm