Learn More
Electron Microscopy Sciences EMS High Alloy DX Biological Tweezers Style 4, 110 mm OAL, 0.06 mm Tip width
SDP
Catalog No. 501930611
Encompass_Preferred
The high alloy biological tweezers are anti-acid and anti-magnetic. The handles are serrated. Tips: Straight, extra fine, superior finish.
OAL: 110 mm
Tip width: 0.06 mm
Tip thickness: 0.03 mm
Catalog No. 50-193-0611 Supplier Electron Microscopy Sciences Supplier No. 785124DX
The high alloy biological tweezers are anti-acid and anti-magnetic. The handles are serrated. Tips: Straight, extra fine, superior finish.
OAL: 110 mm
Tip width: 0.06 mm
Tip thickness: 0.03 mm